Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lauryl acetate
Synonym(s):
Dodecyl acetate, Lauryl acetate
Linear Formula:
CH
3
CO
2
(CH
2
)
11
CH
3
CAS Number:
112-66-3
Molecular Weight:
228.37
EC Number:
203-995-1
Products
Product Category
aroma chemicals (1)
Brand
Sigma-Aldrich (1)
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Food Grade (1)
Halal (1)
Kosher (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
W261602
Lauryl acetate,
≥98%, FG
Expand
View Pricing