![L-Arabinitol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/250/691/de198994-93b8-4886-9647-d5c5815451e9/640/de198994-93b8-4886-9647-d5c5815451e9.png)
L-Arabinitol
![L-Arabinitol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/250/691/de198994-93b8-4886-9647-d5c5815451e9/640/de198994-93b8-4886-9647-d5c5815451e9.png)
Synonym(s):
L-(−)-Arabitol, L-Arabinitol
Empirical Formula (Hill Notation):
C5H12O5
CAS Number:
Molecular Weight:
152.15
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Technique