Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lanthanum(III) chloride
Synonym(s):
Lanthanum trichloride
Linear Formula:
LaCl
3
CAS Number:
10099-58-8
Molecular Weight:
245.26
EC Number:
233-237-5
Products
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Product Line
Purity
Reagent Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
449830
Lanthanum(III) chloride,
AnhydroBeads
™
, −10 mesh, ≥99.99% trace metals basis
Expand
View Pricing
298182
Lanthanum(III) chloride,
AnhydroBeads
™
, −10 mesh, 99.9% trace metals basis
Expand
View Pricing