Skip to Content
Merck
Lanthanum foil, 50x50mm, thickness 0.125mm, as rolled, 99%

Lanthanum

Lanthanum foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Synonym(s):
Lanthanum
Empirical Formula (Hill Notation):
La
CAS Number:
Molecular Weight:
138.91

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Diameter (mm)
Length (mm)
Element
Reagent Type
Core
Showing 1-17 of 17
Compare
Product Number
Description
Pricing