Skip to Content
Merck
Labetalol hydrochloride European Pharmacopoeia (EP) Reference Standard

Labetalol hydrochloride

Labetalol hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzamide hydrochloride, Labetalol hydrochloride
Empirical Formula (Hill Notation):
C19H24N2O3 · HCl
CAS Number:
Molecular Weight:
364.87

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique