Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Krypton-
82
Kr
Linear Formula:
82
Kr
CAS Number:
14191-81-2
Molecular Weight:
81.91
Products
Product Category
gases (5)
stable isotopes (5)
Brand
Sigma-Aldrich (5)
Formula Weight
Physical Form
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
601942
Krypton-
82
Kr,
99.5 atom %
Expand
View Pricing
601926
Krypton-
82
Kr,
99 atom %
Expand
View Pricing
601896
Krypton-
82
Kr,
40 atom %
Expand
View Pricing
601918
Krypton-
82
Kr,
70 atom %
Expand
View Pricing
601934
Krypton-
82
Kr,
90 atom %
Expand
View Pricing