Skip to Content
Merck
KiCqStart® Probe qPCR ReadyMix™, ROX™ with ROX for ABI instruments

KiCqStart® Probe qPCR ReadyMix, ROX

KiCqStart® Probe qPCR ReadyMix™, ROX™ with ROX for ABI instruments
Synonym(s):
Kicqstart® Probe qPCR ReadyMix

Products

Product Category
Brand
Color
Physical Form
Number of Reactions
dNTPs
Hot Start
Sample Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing