![Isovitexin primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/415/974/baf46db0-697c-4059-8560-455c2dbb022e/640/baf46db0-697c-4059-8560-455c2dbb022e.png)
Isovitexin
![Isovitexin primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/415/974/baf46db0-697c-4059-8560-455c2dbb022e/640/baf46db0-697c-4059-8560-455c2dbb022e.png)
Synonym(s):
6-C-Glucosylapigenin, Homovitexin, Saponaretin
Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4