Skip to Content
Merck
Isotretinoin Pharmaceutical Secondary Standard; Certified Reference Material

Isotretinoin

Isotretinoin Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
13-cis-Retinoic acid, Isotretinoin
Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing