Skip to Content
Merck
Isopropyl acetate anhydrous, ZerO2®, ≥99.6%

Isopropyl acetate

Isopropyl acetate anhydrous, ZerO2®, ≥99.6%
Synonym(s):
Acetic acid isopropyl ester, Isopropyl acetate
Linear Formula:
CH3COOCH(CH3)2
CAS Number:
Molecular Weight:
102.13
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique