Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iron(III) perchlorate hydrate
Synonym(s):
Perchloric acid iron(III) salt hydrate
Linear Formula:
Fe(ClO
4
)
3
· xH
2
O
CAS Number:
15201-61-3
Molecular Weight:
354.20 (anhydrous basis)
EC Number:
236-908-0
Products
Color
Formula Weight
Physical Form
Reagent Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
309281
Iron(III) perchlorate hydrate,
crystalline
Expand
View Pricing
326348
Iron(III) perchlorate hydrate,
crystalline, low chloride
Expand
View Pricing