Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1
Products
Manufacturer
Physical Form
Filter Products
Showing 1-20 of 38
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF24979731
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 50 x 50mm, 0.004mm thickness, as rolled, not light tested (nlt)
Expand
View Pricing
GF00407955
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 25 x 25mm, 0.004mm thickness, as rolled, not light tested (nlt)
Expand
View Pricing
GF88627045
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 6mm disks, 0.008mm thickness, as rolled
Expand
View Pricing
GF29509016
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 4mm disks, 0.008mm thickness, as rolled
Expand
View Pricing
GF85307216
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 6mm disks, 0.007mm thickness, as rolled
Expand
View Pricing
GF04318826
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 8mm disks, 0.005mm thickness, as rolled
Expand
View Pricing
GF82854748
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 25 x 25mm, 0.005mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF71314769
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 25 x 25mm, 0.004mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF20360281
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 25 x 25mm, 0.008mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF60929309
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 15mm disks, 0.007mm thickness, as rolled
Expand
View Pricing
GF23882452
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 4mm disks, 0.007mm thickness, as rolled
Expand
View Pricing
GF32991059
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 50 x 50mm, 0.007mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF96106079
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 10mm disks, 0.006mm thickness, as rolled
Expand
View Pricing
GF55168847
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 8mm disks, 0.006mm thickness, as rolled
Expand
View Pricing
GF56843725
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 33 x 50mm, 0.005mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF13765822
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 25 x 25mm, 0.003mm thickness, as rolled, not light tested (nlt)
Expand
View Pricing
GF90902268
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 15mm disks, 0.008mm thickness, as rolled
Expand
View Pricing
GF97527213
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 10mm disks, 0.008mm thickness, as rolled
Expand
View Pricing
GF63402296
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 8mm disks, 0.007mm thickness, as rolled
Expand
View Pricing
GF81063929
Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1,
foil, 25 x 25mm, 0.007mm thickness, as rolled, light tight (lt)
Expand
View Pricing
Page 1 of 2
Page 1 of 2