Irinotecan Related Compound B
Synonym(s):
(4S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3′,4′:6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)dione
Empirical Formula (Hill Notation):
C22H20N2O5
CAS Number:
Molecular Weight:
392.40
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing