![Iopromide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/370/817/31f2b544-f4fe-4f75-a27d-8bbc78c96435/640/31f2b544-f4fe-4f75-a27d-8bbc78c96435.png)
Iopromide
![Iopromide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/370/817/31f2b544-f4fe-4f75-a27d-8bbc78c96435/640/31f2b544-f4fe-4f75-a27d-8bbc78c96435.png)
Empirical Formula (Hill Notation):
C18H24I3N3O8
CAS Number:
Molecular Weight:
791.11
Products
Brand
Formula Weight
Manufacturer
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3