Skip to Content
Merck
Insulin from porcine pancreas powder, ≥27 USP units/mg

Insulin from porcine pancreas

Insulin from porcine pancreas powder, ≥27 USP units/mg
Synonym(s):
Insulin from hog pancreas
Empirical Formula (Hill Notation):
C256H381N65O76S6
CAS Number:
Molecular Weight:
5777.54
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Tissue
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing