IL-3 from mouse
Synonym(s):
HCGF P-cell stimulation factorIL-3 MGC79398, Interleukin-3, MCGF (Mast cell growth factor)Multi-CSF, MGC79399
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Recombinant Host
Showing 1-1 of 1
Compare
Product Number
Description
Pricing