Hypophyllanthin
Synonym(s):
NSC 619044
Empirical Formula (Hill Notation):
C24H30O7
CAS Number:
Molecular Weight:
430.49
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing