Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hypericin
Synonym(s):
4,5,7,4′,5′,7′-Hexahydroxy-2,2′-dimethylnaphthodianthrone, Hypericin
Empirical Formula (Hill Notation):
C
30
H
16
O
8
CAS Number:
548-04-9
Molecular Weight:
504.44
EC Number:
208-941-0
Products
Product Category
bioactive small molecules (1)
Brand
Millipore (1)
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
analytical standard (1)
primary reference standard (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
PHL89226
Hypericin,
phyproof
®
Reference Substance
Expand
View Pricing
400076-M
Hypericin,
Polycyclic dione that has anti-retroviral activity.
Expand
View Pricing
00190585
Hypericin,
primary reference standard
Expand
View Pricing