Skip to Content
Merck
Hydroxyurea United States Pharmacopeia (USP) Reference Standard

Hydroxyurea

Hydroxyurea United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Hydroxycarbamide, Hydroxyurea
Linear Formula:
NH2CONHOH
CAS Number:
Molecular Weight:
76.05

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade