Hydrocortisone 21-acetate
Synonym(s):
11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-acetate, 17-α-Hydroxycorticosterone acetate, 17-Hydroxycorticosterone 21-acetate, 21-Acetoxy-4-pregnene-11β,17α-diol-3,20-dione, 4-Pregnene-11β,17α,21-triol-3,20-dione 21-acetate
Empirical Formula (Hill Notation):
C23H32O6
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Agency
Research Area
Sterility
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing