
Hydramethylnon

Synonym(s):
1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one 2-(1,4,5,6-tetrahydro-5,5-dimethyl-2-pyrimidinyl)hydrazone, NSC 379665, Tetrahydro-5,5-dimethyl-2(1H)-pyrimidinone[3-[4-(trifluoromethyl)phenyl]-1-[2-[4-(trifluoromethyl)phenyl]ethenyl]-2-propylidene]hydrazone
Empirical Formula (Hill Notation):
C25H24F6N4
Products
Product Category
Brand
Formula Weight
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing