Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
HybridSPE
®
-PLus 96-Well solid phase extraction (SPE) Plate
Synonym(s):
HybridSPE
®
-PLus
Products
Product Category
plates (2)
Brand
Supelco (1)
Product Line
Chemical Composition
Volume (ml)
Quality Segment
Package Size
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
575659-U
HybridSPE
®
-PLus 96-Well solid phase extraction (SPE) Plate,
Volume 2 mL, bed wt., 50 mg, pk of 1 ea
Expand
View Pricing
575673-U
HybridSPE
®
-PLus 96-Well solid phase extraction (SPE) Plate,
Volume 2 mL, bed wt., 50 mg, pk of 20 ea
Expand
View Pricing