Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
HPTLC plates, Silica gel 60
Products
Product Category
plates (3)
Brand
Supelco (3)
Length (cm)
Quality Segment
Fluorescence
Matrix Material
Support Material
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.05641
HPTLC plates, Silica gel 60,
pkg of 50 plates, plate L × W 20 cm × 10 cm, glass support
Expand
View Pricing
1.05631
HPTLC plates, Silica gel 60,
pkg of 25 plates, plate L × W 10 cm × 10 cm, glass support
Expand
View Pricing
1.05633
HPTLC plates, Silica gel 60,
pkg of 100 plates, plate L × W 10 cm × 10 cm, glass support
Expand
View Pricing