Skip to Content
Merck
Haloperidol Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material

Haloperidol Related Compound A

Haloperidol Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-{4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidinyl]pphenyl}-1-butanon, 4,4′-Bis[4-(p-chlorophenyl)-4-hydroxypiperidino]butyrophenone
Empirical Formula (Hill Notation):
C32H36Cl2N2O3
CAS Number:
Molecular Weight:
567.55

Products

Brand
Formula Weight
Physical Form
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1