Skip to Content
Merck
vibrant-m

Granisetron Related Compound B

vibrant-m
Synonym(s):
N-[(1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide
Empirical Formula (Hill Notation):
C17H22N4O
CAS Number:
Molecular Weight:
298.38

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1