![Gold nano-urchins 70 nm, maleimide functionalized, conjugation kit](/deepweb/assets/sigmaaldrich/product/images/239/036/e1caaf25-9be1-434a-88a1-5283ba5d621b/640/e1caaf25-9be1-434a-88a1-5283ba5d621b.jpg)
Gold nano-urchins
![Gold nano-urchins 70 nm, maleimide functionalized, conjugation kit](/deepweb/assets/sigmaaldrich/product/images/239/036/e1caaf25-9be1-434a-88a1-5283ba5d621b/640/e1caaf25-9be1-434a-88a1-5283ba5d621b.jpg)
Synonym(s):
Gold nanoparticles, Non-functionalized gold nanoparticles, Gold nano-urchins, Au nanoparticles, Urchins
Products
Brand
pH Value
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-6 of 6
Compare
Product Number
Description
Pricing