Skip to Content
Merck
γ-Nonanoic lactone 97%

γ-Nonanoic lactone

γ-Nonanoic lactone 97%
Synonym(s):
Coconut aldehyde
Empirical Formula (Hill Notation):
C9H16O2
CAS Number:
Molecular Weight:
156.22
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing