Skip to Content
Merck
Glycine sodium salt hydrate ≥99% (HPLC)

Glycine sodium salt hydrate

Glycine sodium salt hydrate ≥99% (HPLC)
Linear Formula:
H2NCH2CO2Na · xH2O
CAS Number:
Molecular Weight:
97.05 (anhydrous basis)

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Amino Acid
Reaction Type
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing