Skip to Content
Merck
Glycine tested according to Ph. Eur.

Glycine

Glycine tested according to Ph. Eur.
Synonym(s):
Aminoacetic acid, Aminoethanoic acid, Glycocoll, Glycine
Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Amino Acid
API Family
Reference Material Type
Special Grade
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2