Skip to Content
Merck
Glyceryl trilinolenate ≥97% (TLC), liquid

Glyceryl trilinolenate

Glyceryl trilinolenate ≥97% (TLC), liquid
Synonym(s):
1,2,3-Tri-(cis,cis,cis-9,12,15-octadecatrienoyl)glycerol, 1,2,3-Trilinolenoylglycerol, Glycerol trilinolenate, Trilinolenin
Empirical Formula (Hill Notation):
C57H92O6
CAS Number:
Molecular Weight:
873.34
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Tissue
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing