Skip to Content
Merck
vibrant-m

Glyceryl monocaprylate, Type II

vibrant-m
Synonym(s):
Glycerol monocaprylate
Empirical Formula (Hill Notation):
C11H22O4
Molecular Weight:
218.29

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1