Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gibberellic acid potassium salt
Synonym(s):
GA
3
, Gibberellin A
3
Empirical Formula (Hill Notation):
C
19
H
21
KO
6
CAS Number:
125-67-7
Molecular Weight:
384.46
EC Number:
204-749-6
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Product Line
Purity
Chemical Composition
Application
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
G1025
Gibberellic acid potassium salt,
suitable for plant cell culture, BioReagent, ~95%, ≥50% total GA
3
basis
Expand
View Pricing