Skip to Content
Merck
Geraniol analytical standard

Geraniol

Geraniol analytical standard
Synonym(s):
trans-3,7-Dimethyl-2,6-octadien-1-ol
Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing