![Geldanamycin from Streptomyces hygroscopicus ≥98% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/672/345/fdac8581-9393-4c6b-af9c-ba50d9efb831/640/fdac8581-9393-4c6b-af9c-ba50d9efb831.png)
Geldanamycin from Streptomyces hygroscopicus
![Geldanamycin from Streptomyces hygroscopicus ≥98% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/672/345/fdac8581-9393-4c6b-af9c-ba50d9efb831/640/fdac8581-9393-4c6b-af9c-ba50d9efb831.png)
Empirical Formula (Hill Notation):
C29H40N2O9
CAS Number:
Molecular Weight:
560.64
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing