Skip to Content
Merck
vibrant-m

Gadodiamide

vibrant-m
Synonym(s):
[5,8-Bis(carboxymethyl)-11-[2-(methylamino)-2-oxoethyl]-3-oxo-2,5,8,11-tetraazatridecan-13-oato (3-)]gadolinium, [N,N-Bis[2-[(carboxymethyl)](methylcarbamoyl)methyl]-amino]ethyl]glycinato(3-)]gadolinium
Empirical Formula (Hill Notation):
C16H26GdN5O8
CAS Number:
Molecular Weight:
573.66

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade