Skip to Content
Merck
Furfuryl mercaptan natural, 98%, FG

Furfuryl mercaptan

Furfuryl mercaptan natural, 98%, FG
Synonym(s):
2-Furanmethanethiol, 2-Furfurylthiol, 2-Furylmethanethiol, Furfuryl mercaptan
Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing