Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Frozen Human Normal CD34+ Hematopoietic Stem Cells
Products
Brand
Sigma-Aldrich (10)
Biological Source
Manufacturer
Physical Form
Tissue
Quality Segment
Package Size
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
HUMANCD34-0002042
Frozen Human Normal CD34+ Hematopoietic Stem Cells,
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Expand
View Pricing
HUMANHL34-0002232
Frozen Human Normal CD34+ Hematopoietic Stem Cells,
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Expand
View Pricing
HUMANHL34-0002235
Frozen Human Normal CD34+ Hematopoietic Stem Cells,
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Expand
View Pricing
HUMANCD34-0002046
Frozen Human Normal CD34+ Hematopoietic Stem Cells,
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Expand
View Pricing
HUMANHL34-0002234
Frozen Human Normal CD34+ Hematopoietic Stem Cells,
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Expand
View Pricing
HUMANCD34-0002044
Frozen Human Normal CD34+ Hematopoietic Stem Cells,
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Expand
View Pricing
HUMANCD34-0002045
Frozen Human Normal CD34+ Hematopoietic Stem Cells,
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Expand
View Pricing
HUMANCD34-0002043
Frozen Human Normal CD34+ Hematopoietic Stem Cells,
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Expand
View Pricing
HUMANHL34-0002236
Frozen Human Normal CD34+ Hematopoietic Stem Cells,
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Expand
View Pricing
HUMANHL34-0002233
Frozen Human Normal CD34+ Hematopoietic Stem Cells,
human peripheral blood (CD34+ Hematopoietic Stem Cells)
Expand
View Pricing