Fondaparinux sodium identification
Synonym(s):
Fondaparinux sodium, Fondaparin sodium, Methyl O-2-deoxy-6-O-sulfo-2-(sulfoamino)-α-D-glucopyranosyl-(14)-O-β-D-glucopyranuronosyl-(14)-O-2-deoxy-3,6-di-O-sulfo-2-(sulfoamino)-α-D-glucopyranosyl-(14)-O-2-O-sulfo-α-L-idopyranuronosyl-(14)-2-deoxy-2-(sulfoamino)-α-D-glucopyranoside 6-(hydrogen sulfate) sodium salt, SR-90107A
Empirical Formula (Hill Notation):
C31H43N3O49S8 · 10Na
CAS Number:
Molecular Weight:
1728.08
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing