![Flurbiprofen Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material, certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/133/823/7b03bb2a-87a8-4889-a336-07cd844fe413/640/7b03bb2a-87a8-4889-a336-07cd844fe413.jpg)
Flurbiprofen Related Compound A
![Flurbiprofen Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material, certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/133/823/7b03bb2a-87a8-4889-a336-07cd844fe413/640/7b03bb2a-87a8-4889-a336-07cd844fe413.jpg)
Synonym(s):
Biprofen, 2-(4-Biphenylyl)propionic acid, NSC 16300
Empirical Formula (Hill Notation):
C15H14O2
CAS Number:
Molecular Weight:
226.27
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2