Skip to Content
Merck
Flucycloxuron PESTANAL®, analytical standard, mixture of (E) and (Z) isomers

Flucycloxuron

Flucycloxuron PESTANAL®, analytical standard, mixture of (E) and (Z) isomers
Synonym(s):
N-[[[4-[[[[(4-Chlorophenyl)cyclopropylmethylene]amino]oxy]methyl]phenyl]amino]carbonyl]-2,6-difluorobenzamide
Empirical Formula (Hill Notation):
C25H20ClF2N3O3
CAS Number:
Molecular Weight:
483.89

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing