
Ferroin indicator solution

Synonym(s):
1,10-Phenanthroline iron(II) sulfate complex, o-Phenanthroline ferrous sulfate complex
Linear Formula:
[Fe(C12H8N2)3]SO4
CAS Number:
Molecular Weight:
692.52
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1