
Ferrocenecarboxaldehyde

Synonym(s):
Cyclopentadienyl(formylcyclopentadienyl)iron
Empirical Formula (Hill Notation):
C11H10FeO
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Reagent Type
Core
Showing 1-1 of 1