
Fenoxaprop-P-ethyl

Synonym(s):
Ethyl (R)-2-{4-[(6-chlorobenzoxazol-2-yl)oxy]phenoxy}propionate
Empirical Formula (Hill Notation):
C18H16ClNO5
CAS Number:
Molecular Weight:
361.78
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2