![Fenoterol for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/226/379/37d5dfb7-2c64-46b2-b61b-8dff1f843527/640/37d5dfb7-2c64-46b2-b61b-8dff1f843527.png)
Fenoterol for system suitability
![Fenoterol for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/226/379/37d5dfb7-2c64-46b2-b61b-8dff1f843527/640/37d5dfb7-2c64-46b2-b61b-8dff1f843527.png)
Synonym(s):
Fenoterol hydrobromide, 2-(3,5-Dihydroxyphenyl)-2-hydroxy-2′-(4-hydroxyphenyl)-1′-methyldiethylamine hydrobromide
Empirical Formula (Hill Notation):
C17H21NO4 · HBr
CAS Number:
Molecular Weight:
384.26
Products
Formula Weight
Gene Alias
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing