Fenofibrate Related Compound A
Synonym(s):
4-Chloro-4′-hydroxybenzophenone, (4-Chlorophenyl)(4-hydroxyphenyl)methanone
Linear Formula:
ClC6H4COC6H4OH
CAS Number:
Molecular Weight:
232.66
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2