Felbinac
Synonym(s):
4-Biphenylacetic acid
Linear Formula:
C6H5C6H4CH2CO2H
CAS Number:
Molecular Weight:
212.24
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing