![Famciclovir Related Compound F pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/357/868/d6990400-57a0-4d47-a554-ccc0041e9d3e/640/d6990400-57a0-4d47-a554-ccc0041e9d3e.png)
Famciclovir Related Compound F
![Famciclovir Related Compound F pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/357/868/d6990400-57a0-4d47-a554-ccc0041e9d3e/640/d6990400-57a0-4d47-a554-ccc0041e9d3e.png)
Synonym(s):
2-Amino-6-chloropurine, 6-Chloro-2-purinamine, 6-Chloroguanine
Empirical Formula (Hill Notation):
C5H4ClN5
CAS Number:
Molecular Weight:
169.57
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2