Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5
Products
Manufacturer
Physical Form
Filter Products
Showing 1-20 of 26
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF04488082
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 25 x 25mm, 0.02mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF53085203
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 25 x 25mm, 0.025mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF26346337
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 100 x 100mm, 0.02mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF61010772
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 50 x 50mm, 0.02mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF06180982
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 25mm disks, 0.025mm thickness, annealed
Expand
View Pricing
GF99535832
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 0.5m coil, 0.025mm thickness, annealed
Expand
View Pricing
GF35650201
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 50 x 50mm, 0.025mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF31249447
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 15mm disks, 0.025mm thickness, annealed
Expand
View Pricing
GF28859596
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 8mm disks, 0.025mm thickness, annealed
Expand
View Pricing
GF84872122
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 8mm disks, 0.02mm thickness, annealed
Expand
View Pricing
GF93291835
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 4mm disks, 0.02mm thickness, annealed
Expand
View Pricing
GF41979358
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 1m coil, 0.025mm thickness, annealed
Expand
View Pricing
GF56275173
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 100 x 100mm, 0.025mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF27005970
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 15mm disks, 0.02mm thickness, annealed
Expand
View Pricing
GF09016040
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 10mm disks, 0.02mm thickness, annealed
Expand
View Pricing
GF61478972
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 6mm disks, 0.02mm thickness, annealed
Expand
View Pricing
GF07449992
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 10mm disks, 0.025mm thickness, annealed
Expand
View Pricing
GF27430871
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 6mm disks, 0.025mm thickness, annealed
Expand
View Pricing
GF47419443
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 4mm disks, 0.025mm thickness, annealed
Expand
View Pricing
GF68424688
Evanohm
®
- precision resistance alloy, Ni75Cr20Al2.5Cu2.5,
foil, 50mm disks, 0.02mm thickness, annealed
Expand
View Pricing
Page 1 of 2
Page 1 of 2