Ethylparaben Sodium
Synonym(s):
Ethyl 4-hydroxybenzoate sodium salt
Empirical Formula (Hill Notation):
C9H9NaO3
CAS Number:
Molecular Weight:
188.16
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1