Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ethyl gallate
Linear Formula:
3,4,5-(OH)
3
C
6
H
2
COOC
2
H
5
CAS Number:
831-61-8
Molecular Weight:
198.17
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Purum (1)
analytical standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
48640
Ethyl gallate,
antioxidant, ≥96.0% (HPLC)
Expand
View Pricing
PHL83080
Ethyl gallate,
phyproof
®
Reference Substance
Expand
View Pricing